Learn More
ABclonal Technology GLI1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A146751000UL
Encompass_Preferred
This gene encodes a member of the Kruppel family of zinc finger proteins. The encoded transcription factor is activated by the sonic hedgehog signal transduction cascade and regulates stem cell proliferation. The activity and nuclear localization of this protein is negatively regulated by p53 in an inhibitory loop. Multiple transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-06833