Learn More
Aviva Systems Biology Corp GLI2 Antibody - middle region (ARP31885_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31885_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GLI2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human GLI2. Alternate names: HPE9, THP1, THP2ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-52841