Learn More
Aviva Systems Biology Corp GLI4 Antibody - N-terminal region (ARP36136_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36136_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GLI4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GLI4. Alternate names: HKR4, ZNF928ApplicationWB. Species reactivity: Human
Catalog No. 50-014-63080