Learn More
Abcam Glibenclamide (Glyburide), K+ channel blocker, 100MG
Catalog No. 502707734
Encompass_Preferred
MW 494 Da, Purity >99%. Selective blocker of ATP-sensitive (KIR6.x) inward rectifier K+ channels. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7734 Supplier Abcam Supplier No. AB120267100MG
MW 494 Da, Purity >99%. Selective blocker of ATP-sensitive (KIR6.x) inward rectifier K+ channels. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement