Learn More
Medchemexpress LLC Gliclazide-d4 | 1185039-30-8 | 99.9% | 5 MG
SDP
Catalog No. 5000405325
Encompass_Preferred
Gliclazide-d4 is the deuterium-labeled form of Gliclazide, an antidiabetic agent that functions as a whole-cell beta-cell ATP-sensitive potassium currents blocker with an IC50 of 184 nM. Deuteration, the process of incorporating stable heavy isotopes like hydrogen and carbon into drug molecules, is used for quantitation during drug development and has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules.
  • Largely used as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-05325 Supplier Medchemexpress LLC Supplier No. HYB0753S5MG
Gliclazide-d4 is the deuterium-labeled form of Gliclazide, an antidiabetic agent that functions as a whole-cell beta-cell ATP-sensitive potassium currents blocker with an IC50 of 184 nM. Deuteration, the process of incorporating stable heavy isotopes like hydrogen and carbon into drug molecules, is used for quantitation during drug development and has the potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules.
  • Largely used as tracers for quantitation during the drug development process.
  • Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.