Learn More
Cayman Chemical GlIclazIde-d4 1mg
SDP
Supplier:  Cayman Chemical 336101 MG
Encompass_Preferred
An internal standard for the quantification of gliclazide by GC- or LC-MS
Catalog No. 50-295-0602