Learn More
Cayman Chemical Gliclazide-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1185039-30-8, 5 mg
SDP
Catalog No. 502950185
Encompass_Preferred
An internal standard for the quantification of gliclazide by GC- or LC-MS
Catalog No. 50-295-0185 Supplier Cayman Chemical Supplier No. 336105 MG
An internal standard for the quantification of gliclazide by GC- or LC-MS