Learn More
AdipoGen Glimepiride (1 g)
SDP
Catalog No. 502054466
Encompass_Preferred
Chemical. CAS: 93479-97-1. Formula: C24H34N4O5S. MW: 490.62. Glimepiride is a long-acting sulfonylurea antidiabetic agent inhibiting ATP-sensitive potassium (KATP) channels in pancreatic beta-cells, which leads to the release of insulin. In addition, Glimepiride increases the activity of intracellular insulin receptors. Studies conducted on adipocytes and skeletal muscle suggest that Glimepiride induces the PI3 kinase (PI3K) and Akt pathway, along with insulin receptor substrate-1/2 and endothelial nitric oxide synthase and stimulates glucose transporter 1 and 4 (GLUT1/4) expression. Glimepiride also increases osteoblast proliferation and differentiation, which is thought to be related to its ability to activate the PI3K and Akt pathway and exhibits neuroprotective benefit, decreasing expression and activity of BACE1 and amyloid-beta (Abeta) in neurons in a PPARgamma-dependent manner.
Catalog No. 50-205-4466 Supplier AdipoGen Supplier No. CDXG0208G001
Chemical. CAS: 93479-97-1. Formula: C24H34N4O5S. MW: 490.62. Glimepiride is a long-acting sulfonylurea antidiabetic agent inhibiting ATP-sensitive potassium (KATP) channels in pancreatic beta-cells, which leads to the release of insulin. In addition, Glimepiride increases the activity of intracellular insulin receptors. Studies conducted on adipocytes and skeletal muscle suggest that Glimepiride induces the PI3 kinase (PI3K) and Akt pathway, along with insulin receptor substrate-1/2 and endothelial nitric oxide synthase and stimulates glucose transporter 1 and 4 (GLUT1/4) expression. Glimepiride also increases osteoblast proliferation and differentiation, which is thought to be related to its ability to activate the PI3K and Akt pathway and exhibits neuroprotective benefit, decreasing expression and activity of BACE1 and amyloid-beta (Abeta) in neurons in a PPARgamma-dependent manner.