Learn More
Medchemexpress LLC Glimepiride | 93479-97-1 | 99.9% | 490.62 g/mol | C24H34N4O5S | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0104R10MG
Encompass_Preferred
Glimepiride (standard) is an analytical reference standard of the sulfonylurea antidiabetic compound glimepiride intended for research and analytical and method development applications. Supplied as a 10 mg pack, the certificate of analysis reports an LCMS purity of 99.92%. Molecular formula C24H34N4O5S; molecular weight 490.62 g/mol. Not for human or veterinary use.
  • Analytical standard for method development and quality control
  • High reported purity by LCMS (99.92%)
  • Compact 10 MG pack suitable for laboratory testing
  • Molecular weight 490.62 g/mol for reference calculations
  • Intended for research use only, not for clinical use
Catalog No. 50-002-98941