Learn More
Medchemexpress LLC Glimepiride | 93479-97-1 | 99.9% | C24H34N4O5S | 5 G
Supplier: Medchemexpress LLC HYB01045G
Glimepiride is a medium-to-long acting sulfonylurea anti-diabetic compound. It has shown to lower blood glucose more effectively than glibenclamide and may decrease extracellular Aβ40 and Aβ42 levels, suggesting its potential for Alzheimer's disease associated with diabetes.
- Medium-to-long acting sulfonylurea anti-diabetic compound.
- Lower risk of hypoglycemia and less weight gain compared to other sulfonylureas.
- May be safer for patients with cardiovascular disease due to a lack of detrimental effects on ischemic preconditioning.
- Potential for Alzheimer's disease associated with diabetes by decreasing extracellular Aβ40 and Aβ42 levels.