Learn More
Medchemexpress LLC Glimepiride | 93479-97-1 | 99.9% | C24H34N4O5S | 500 MG
SDP
Catalog No. 5000432830
Encompass_Preferred
Glimepiride (Glimperide) is a medium-to-long acting sulfonylurea anti-diabetic compound. It acts mainly by stimulating insulin release from pancreatic beta cells and increasing the sensitivity to insulin at receptor sites, thereby lowering blood sugar. Glimepiride is effective in controlling blood glucose and is often used as an adjunct to diet and exercise to improve glycemic control in adults with type 2 diabetes mellitus.
  • Medium-to-long acting anti-diabetic compound
  • Stimulates insulin release from pancreatic beta cells
  • Increases insulin sensitivity at receptor sites
  • Effective in controlling blood glucose
  • Used to improve glycemic control in type 2 diabetes mellitus
Catalog No. 50-004-32830 Supplier Medchemexpress LLC Supplier No. HYB0104500MG
Glimepiride (Glimperide) is a medium-to-long acting sulfonylurea anti-diabetic compound. It acts mainly by stimulating insulin release from pancreatic beta cells and increasing the sensitivity to insulin at receptor sites, thereby lowering blood sugar. Glimepiride is effective in controlling blood glucose and is often used as an adjunct to diet and exercise to improve glycemic control in adults with type 2 diabetes mellitus.
  • Medium-to-long acting anti-diabetic compound
  • Stimulates insulin release from pancreatic beta cells
  • Increases insulin sensitivity at receptor sites
  • Effective in controlling blood glucose
  • Used to improve glycemic control in type 2 diabetes mellitus