Learn More
Cayman Chemical Glimepiride-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 2699607-96-8, 1 mg
SDP
Catalog No. 502939267
Encompass_Preferred
An internal standard for the quantification of glimepiride by GC- or LC-MS
Catalog No. 50-293-9267 Supplier Cayman Chemical Supplier No. 278461 MG
An internal standard for the quantification of glimepiride by GC- or LC-MS