Learn More
Medchemexpress LLC Glimepiride (Standard) | 93479-97-1 | >99.9% | C24H34N4O5S | 100 MG
SDP
Catalog No. 5000432434
Encompass_Preferred
Glimepiride (Standard) is an analytical standard of Glimepiride, a medium-to-long acting sulfonylurea anti-diabetic compound. It is intended for research and analytical applications, and is not for patient use.
  • Medium-to-long acting sulfonylurea anti-diabetic compound
  • Lowers blood glucose
  • Attenuates Aβ production by suppressing BACE1 activity
  • Associated with lower risk of hypoglycemia
  • Less weight gain compared to other sulfonylureas
  • May be safer for patients with cardiovascular disease
Catalog No. 50-004-32434 Supplier Medchemexpress LLC Supplier No. HYB0104R100MG
Glimepiride (Standard) is an analytical standard of Glimepiride, a medium-to-long acting sulfonylurea anti-diabetic compound. It is intended for research and analytical applications, and is not for patient use.
  • Medium-to-long acting sulfonylurea anti-diabetic compound
  • Lowers blood glucose
  • Attenuates Aβ production by suppressing BACE1 activity
  • Associated with lower risk of hypoglycemia
  • Less weight gain compared to other sulfonylureas
  • May be safer for patients with cardiovascular disease