Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Glimepiride sulfonamide | 119018-29-0 | MFCD02955393 | 98.0% | 351.43 g/mol | C16H21N3O4S | 100 MG
SDP

Catalog No. 5000209164
Encompass_Preferred

Glimepiride sulfonamide is a chemical intermediate used in the synthesis of the antidiabetic agent glimepiride. It is supplied as an off-white to white solid powder and as DMSO solutions for research and analytical use. The compound has a molecular weight of approximately 351.4 g/mol and chemical formula C16H21N3O4S, and typical analytical standards are supplied at ~98% purity.

  • Used as an intermediate for the synthesis of glimepiride and related compounds.
  • Available as a solid powder and as a DMSO solution (10 mM, 1 mL).
  • Physical form: off-white to white solid powder suitable for synthetic and analytical applications.
  • Molecular weight: ~351.4 g/mol; chemical formula: C16H21N3O4S.
  • Storage: powder at cold temperatures for long-term stability; in solvent, store at ultracold temperatures for short-term stability.
  • Analytical standard available with typical purity around 98%.

Catalog No. 50-002-09164 Supplier Medchemexpress LLC Supplier No. HYW041324100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Glimepiride sulfonamide is a chemical intermediate used in the synthesis of the antidiabetic agent glimepiride. It is supplied as an off-white to white solid powder and as DMSO solutions for research and analytical use. The compound has a molecular weight of approximately 351.4 g/mol and chemical formula C16H21N3O4S, and typical analytical standards are supplied at ~98% purity.

  • Used as an intermediate for the synthesis of glimepiride and related compounds.
  • Available as a solid powder and as a DMSO solution (10 mM, 1 mL).
  • Physical form: off-white to white solid powder suitable for synthetic and analytical applications.
  • Molecular weight: ~351.4 g/mol; chemical formula: C16H21N3O4S.
  • Storage: powder at cold temperatures for long-term stability; in solvent, store at ultracold temperatures for short-term stability.
  • Analytical standard available with typical purity around 98%.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.