Learn More
Sigma Aldrich Fine Chemicals Biosciences Glindemann PTFE sealing rings, 50/pk
Catalog No. NC0948883
Encompass
This narrow ring seal is an alternative to a PTFE joint sleeve. Greaseless protection for air- and moisture- sensitive compounds. Can be used for prolonged storage under vacuum. Slide easily onto medium and full length taper joints.
Catalog No. NC0948883 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5023161PAK
This narrow ring seal is an alternative to a PTFE joint sleeve. Greaseless protection for air- and moisture- sensitive compounds. Can be used for prolonged storage under vacuum. Slide easily onto medium and full length taper joints.