Learn More
Medchemexpress LLC HY-N6727 5mg Medchemexpress, Gliotoxin CAS:67-99-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN67275MG
Encompass_Preferred
Medchemexpress, HY-N6727 5mg Gliotoxin CAS:67-99-2 Gliotoxin is a secondary metabolite, the most abundant mycotoxin secreted by A. fumigatus, inhibits the phagocytosis of macrophages and the immune functions of other immune cells . Gliotoxin inhibits inducible NF-κB activity by preventing IκB degradation, which consequently induces host-cell apoptosis. Gliotoxin activates PKA and increases intracellular cAMP concentration; modulates actin cytoskeleton rearrangement to facilitate A. fumigatus internalization into lung epithelial cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2959