Learn More
Medchemexpress LLC Glipizide-d11 | 1189426-07-0 | 96.4% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0254S5MG
Encompass_Preferred
Glipizide-d11 is the deuterium labeled Glipizide. Glipizide (CP 2872; K 4024) is a potent, orally active, sulfonylurea class anti-diabetic agent used for type 2 diabetes mellitus research, but not for type 1. Glipizide acts by partially blocking ATP-sensitive potassium (KATP) channels among β cells of pancreatic islets of Langerhans.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-06950