Learn More
Cayman Chemical GlIpIzIde-d11 500ug
SDP
Catalog No. 502991788
Encompass_Preferred
An internal standard for the quantification of glipizide by GC- or LC-MS
Catalog No. 50-299-1788 Supplier Cayman Chemical Supplier No. 30074500 UG
An internal standard for the quantification of glipizide by GC- or LC-MS