Learn More
Medchemexpress LLC Glipizide (Standard) | 29094-61-9 | 99.7% | 100 MG
SDP
Catalog No. 5000406983
Encompass_Preferred
Glipizide (Standard) is an analytical standard intended for research and analytical applications. This potent, orally active sulfonylurea class anti-diabetic agent is used for type 2 diabetes mellitus research, acting by partially blocking ATP-sensitive potassium channels among β cells of pancreatic islets of Langerhans.
  • Analytical standard for research and analytical applications
  • Potent, orally active sulfonylurea class anti-diabetic agent
  • Used for type 2 diabetes mellitus research
  • Acts by partially blocking ATP-sensitive potassium (KATP) channels
  • Reference standard supplied assay
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS
Catalog No. 50-004-06983 Supplier Medchemexpress LLC Supplier No. HYB0254R100MG
Glipizide (Standard) is an analytical standard intended for research and analytical applications. This potent, orally active sulfonylurea class anti-diabetic agent is used for type 2 diabetes mellitus research, acting by partially blocking ATP-sensitive potassium channels among β cells of pancreatic islets of Langerhans.
  • Analytical standard for research and analytical applications
  • Potent, orally active sulfonylurea class anti-diabetic agent
  • Used for type 2 diabetes mellitus research
  • Acts by partially blocking ATP-sensitive potassium (KATP) channels
  • Reference standard supplied assay
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS