Learn More
Medchemexpress LLC Glipizide (Standard) | 29094-61-9 | 99.7% | 25 MG
SDP
Catalog No. 5000450581
Encompass_Preferred
Glipizide (Standard) is an analytical standard of Glipizide, intended for research and analytical applications. It is a potent, orally active sulfonylurea class anti-diabetic agent used for type 2 diabetes mellitus research.
  • Potent, orally active sulfonylurea class anti-diabetic agent
  • Used for type 2 diabetes mellitus research
  • Acts by partially blocking ATP-sensitive potassium (KATP) channels
  • Suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • IC50: 6.4 nM ATP-sensitive potassium (KATP) channels in primary mouse pancreatic β cells
Catalog No. 50-004-50581 Supplier Medchemexpress LLC Supplier No. 25MGHYB0254R
Glipizide (Standard) is an analytical standard of Glipizide, intended for research and analytical applications. It is a potent, orally active sulfonylurea class anti-diabetic agent used for type 2 diabetes mellitus research.
  • Potent, orally active sulfonylurea class anti-diabetic agent
  • Used for type 2 diabetes mellitus research
  • Acts by partially blocking ATP-sensitive potassium (KATP) channels
  • Suitable for qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • IC50: 6.4 nM ATP-sensitive potassium (KATP) channels in primary mouse pancreatic β cells