Learn More
Aviva Systems Biology Corp GLIPR1L1 Antibody - middle region (ARP55549_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55549_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GLIPR1L1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GLIPR1L1. Alternate names: ALKN2972, MGC26856, PRO7434ApplicationWB. Species reactivity: Human
Catalog No. 50-286-056