Learn More
Aviva Systems Biology Corp GLIS2 Antibody - N-terminal region (ARP30036_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30036_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GLIS2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GLIS2. Alternate names: NKL, NPHP7ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54106