Learn More
Aviva Systems Biology Corp GLIS3 Antibody - C-terminal region (ARP39925_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39925_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GLIS3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human GLIS3. Alternate names: FLJ38999, FLJ90578, MGC33662, ZNF515ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53162