Learn More
Medchemexpress LLC Gln-AMS TFA | 99.85% | 588.47 | 50 MG
SDP
Catalog No. 5000437902
Encompass_Preferred
Gln-AMS (TFA) is a type Ia aminoacyl-tRNA synthetase (AARS) inhibitor. It specifically inhibits glutaminyl-tRNA synthetase (GlnRS) with a Ki of 1.32 μM. This product is intended for research use only.
  • Inhibits glutaminyl-tRNA synthetase.
  • High purity of 99.85%.
  • Available in various quantities.
  • Data sheet, COA, SDS, and handling instructions provided.
  • Supported by customer reviews and publications.
Catalog No. 50-004-37902 Supplier Medchemexpress LLC Supplier No. HY112861A50MG
Gln-AMS (TFA) is a type Ia aminoacyl-tRNA synthetase (AARS) inhibitor. It specifically inhibits glutaminyl-tRNA synthetase (GlnRS) with a Ki of 1.32 μM. This product is intended for research use only.
  • Inhibits glutaminyl-tRNA synthetase.
  • High purity of 99.85%.
  • Available in various quantities.
  • Data sheet, COA, SDS, and handling instructions provided.
  • Supported by customer reviews and publications.