Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gln-AMS TFA | 99.9% | 588.47 | 100 MG
SDP

Catalog No. 5000437558
Encompass_Preferred

Gln-AMS (TFA) is a type Ia aminoacyl-tRNA synthetase (AARS) inhibitor, specifically inhibiting glutaminyl-tRNA synthetase (GlnRS) with a Ki of 1.32 μM.

  • Inhibits glutaminyl-tRNA synthetase (GlnRS)
  • Type Ia aminoacyl-tRNA synthetase (AARS) inhibitor
  • White to off-white solid
  • Purity of 99.85%
  • Soluble in DMSO at 100 mg/mL
  • Store at -20°C under nitrogen

Catalog No. 50-004-37558 Supplier Medchemexpress LLC Supplier No. HY112861A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gln-AMS (TFA) is a type Ia aminoacyl-tRNA synthetase (AARS) inhibitor, specifically inhibiting glutaminyl-tRNA synthetase (GlnRS) with a Ki of 1.32 μM.

  • Inhibits glutaminyl-tRNA synthetase (GlnRS)
  • Type Ia aminoacyl-tRNA synthetase (AARS) inhibitor
  • White to off-white solid
  • Purity of 99.85%
  • Soluble in DMSO at 100 mg/mL
  • Store at -20°C under nitrogen

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.