Learn More
CPC Scientific H-His-Ser-Leu-Gly-Lys-Gln-Leu-Gly-His-Pro-Asp-Lys-Phe-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PLPS001A
Encompass
SEQUENCE: H-His-Ser-Leu-Gly-Lys-Gln-Leu-Gly-His-Pro-Asp-Lys-Phe-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: HSLGKQLGHPDKFMOLECULAR FORMULA: C66H102N20O18MOLECULAR WEIGHT: 1463.67STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: Multiple SclerosisREFERENCES:Zsolt Ills, et. al. J. Immunol. 175, 1491 ( 2005)Proc. Natl. Acad. Sci. USA 95, 264 (1998)Munder, M. et al. J. Exp. Med. 196, 1151 (2002)Murtaza, A. et al. Int. Immunol. 11, 407 (1999)SKU(s): PLPS-001A
Catalog No. 50-211-0214