Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC [Gln8]-C517 (LH-RH), chicken | 47922-48-5 | 99.97% | 1154.23 | 100 MG
SDP

Catalog No. 5000423926
Encompass_Preferred

[Gln8]-C517 (LH-RH), chicken is an avian hypothalamic peptide that stimulates the release of gonadotropins from the anterior pituitary, thereby regulating reproductive functions.

  • Intended for research use only.
  • Solid appearance.
  • White to off-white color.
  • Sequence: {pGLU}-His-Trp-Ser-Tyr-Gly-Leu-Gln-Pro-Gly.
  • Soluble in H2O at 100 mg/mL (requires ultrasonic).
  • Targets GnRH receptor.
  • Involved in GPCR/G protein pathway.

Catalog No. 50-004-23926 Supplier Medchemexpress LLC Supplier No. HYP1905100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

[Gln8]-C517 (LH-RH), chicken is an avian hypothalamic peptide that stimulates the release of gonadotropins from the anterior pituitary, thereby regulating reproductive functions.

  • Intended for research use only.
  • Solid appearance.
  • White to off-white color.
  • Sequence: {pGLU}-His-Trp-Ser-Tyr-Gly-Leu-Gln-Pro-Gly.
  • Soluble in H2O at 100 mg/mL (requires ultrasonic).
  • Targets GnRH receptor.
  • Involved in GPCR/G protein pathway.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.