Learn More
Glo Germ Company Glo Germ MIST Combination pack
SDP
Catalog No. NC0940521
Encompass_Preferred
Glo Germ MIST Combination pack includes the Glo Germ MIST Spray Canister and an 8 ounce refill. These products together effectively demonstrates airborne microbe, cough and sneeze simulation.
Catalog No. NC0940521 Supplier Glo Germ Company Supplier No. MSTCMB
Glo Germ MIST Combination pack includes the Glo Germ MIST Spray Canister and an 8 ounce refill. These products together effectively demonstrates airborne microbe, cough and sneeze simulation.