Learn More
Echelon Biosciences Research Labs GLO-PI(3,4,5)P3 BODIPYFL100ug
SDP
Catalog No. 50720122
Encompass
BODIPY FL PI(3,4,5)P3 (BODIPY FL Phosphatidylinositol 3,4,5-trisphosphate) is a water soluble analog of PI(3,4,5)P3 (PIP3) labeled with the green fluorophore, Bodipy-FL at the sn-1 position.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 3,4,5-trisphosphate (PIP3), formed from PI(4,5)P2 though phosphorylation by PI 3-kinase, activates numerous signaling pathways resulting in cell proliferation, growth, survival, glucose transport and protein synthesis. High PIP3 levels from disregulation of PI3-K have been demonstrated in cancer and inflammatory diseases. PIP3 is hydrolyzed by the phosphatases PTEN to PI(4,5)P2 and SHIP to PI(3,4)P2. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-122 Supplier Echelon Biosciences Research Labs Supplier No. C39F6A100UG
BODIPY FL PI(3,4,5)P3 (BODIPY FL Phosphatidylinositol 3,4,5-trisphosphate) is a water soluble analog of PI(3,4,5)P3 (PIP3) labeled with the green fluorophore, Bodipy-FL at the sn-1 position.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 3,4,5-trisphosphate (PIP3), formed from PI(4,5)P2 though phosphorylation by PI 3-kinase, activates numerous signaling pathways resulting in cell proliferation, growth, survival, glucose transport and protein synthesis. High PIP3 levels from disregulation of PI3-K have been demonstrated in cancer and inflammatory diseases. PIP3 is hydrolyzed by the phosphatases PTEN to PI(4,5)P2 and SHIP to PI(3,4)P2. This item is non returnable