Learn More
Echelon Biosciences Research Labs GloPIPs BODIPY TMR PI(3)P 100 ug
SDP
Catalog No. 50720051
Encompass
Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 3-phosphate (PI(3)P) is enriched in early endosomes having roles in endosome fusion and receptor sorting and internalization in multivesicular bodies. PI(3)P has also been found at the plasma membrane and is involved in the translocation of the glucose transport protein GLUT4.GloPIPs are labeled at the sn-1 position with BODIPY-FL (505/513), BODIPY-TMR (544/574), or biotin. The sn-1 linkage in these labeled PIPs has been changed to a more stable amide from an ester. Results from experiments conducted at Echelon indicated that this minor chemical change did not have a significant effect in any of the applications tested. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-051 Supplier Echelon Biosciences Research Labs Supplier No. C03M6A100UG
Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 3-phosphate (PI(3)P) is enriched in early endosomes having roles in endosome fusion and receptor sorting and internalization in multivesicular bodies. PI(3)P has also been found at the plasma membrane and is involved in the translocation of the glucose transport protein GLUT4.GloPIPs are labeled at the sn-1 position with BODIPY-FL (505/513), BODIPY-TMR (544/574), or biotin. The sn-1 linkage in these labeled PIPs has been changed to a more stable amide from an ester. Results from experiments conducted at Echelon indicated that this minor chemical change did not have a significant effect in any of the applications tested. This item is non returnable