Learn More
Echelon Biosciences Research Labs GLO-PI(4,5)P2 BODIPYTMRC16 100ug
SDP
Catalog No. 50720141
Encompass
hosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 4,5-bisphosphate (PIP2) has been shown to play a central role in a variety of cellular functions. Amongst its many functions, PIP2 is a substrate for Phospholipase C-coupled G-protein pathways involved in intracellular calcium release in a number of tissues. It is also a substrate for class I phosphoinositide 3-kinase (PI3-K) forming PI(3,4,5)P3. GloPIPs are labeled at the sn-1 position with BODIPY-FL (505/513), BODIPY-TMR (544/574), or biotin. The sn-1 linkage in these labeled PIPs has been changed to a more stable amide from an ester. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-141 Supplier Echelon Biosciences Research Labs Supplier No. C45M16A100UG
hosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 4,5-bisphosphate (PIP2) has been shown to play a central role in a variety of cellular functions. Amongst its many functions, PIP2 is a substrate for Phospholipase C-coupled G-protein pathways involved in intracellular calcium release in a number of tissues. It is also a substrate for class I phosphoinositide 3-kinase (PI3-K) forming PI(3,4,5)P3. GloPIPs are labeled at the sn-1 position with BODIPY-FL (505/513), BODIPY-TMR (544/574), or biotin. The sn-1 linkage in these labeled PIPs has been changed to a more stable amide from an ester. This item is non returnable