Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Staples Advantage Glo–Write™ Fluorescent Markers

Catalog No. 13384
Change view
Click to view available options
Fluorescent Markers
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
13-384 Fluorescent Markers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-384 Supplier Staples Advantage Supplier No. 74194

Includes

Five fluorescent paint markers: pink, yellow, green, light blue and white.

Long-lasting

Bright, colorful markers wipe off easily with water.

Specifications

Product Type Fluorescent Marker
Tip Style Bullet
Color Assorted
Includes Five fluorescent paint markers pink, yellow, green, light blue and white
For Use With (Application) Ideal for signs, posting daily specials and highlighting sales
For Use With (Equipment) Quartet black marker boards
Quantity 5 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.