Learn More
NATtrol™ Global Fever Panel
SDP
Catalog No. 22158470 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
22 x 0.3 mL
Catalog No. Quantity
22-158-470 22 x 0.3 mL
1 options
Catalog No. 22-158-470 Supplier NATtrol™ Supplier No. NATGFPBIO
Add to Cart
Add to Cart
Global Fever Panel is designed to evaluate the performance of nucleic acid tests to determine the presence of bacterial, protozoan and viral nucleic acids. Can also be used for validation of clinical assays, development of diagnostic tests and training of laboratory personnel.
  • Formulated with purified, intact bacterial cells and viral particles
  • Microorganisms have been chemically modified to render them non-infectious and refrigerator stable
  • NATGFP-BIO contains 22 x 0.3 mL vials of bacterial and viral NATtrol™. The panel members are supplied in a proprietary matrix.
Specifications
Manufacturing Origin United States
Product Type Panel
Sterility Non-sterile
Target Organism Class Bacterial Nucleic Acids, Viral Nucleic Acids, Protozoan Nucleic Acids
Test Type Verification Panel
Quantity 22 x 0.3 mL