Learn More
MSC Global Glove Cut Resistant Gloves, 13-Gauge, ANSI A4, 1 Pair
Catalog No. 5000794938
Encompass
These enhanced seamless gloves offer dependable cut resistance with a 13-gauge Tuffalene® UHMWPE shell. A black foam nitrile palm dip provides excellent wet/oily grip and improved abrasion resistance.
  • ANSI level A4 cut resistance
  • EN 388 levels: 4X43D
  • Features a 13-gauge seamless shell
  • Foam nitrile coated palm for high grip
  • Excellent abrasion resistance
  • Knit wrist for secure fit and comfort
  • Suitable for sharp object handling, sheet metal work, and glass handling
Catalog No. 50-007-94938 Supplier MSC Supplier No. 41471277
These enhanced seamless gloves offer dependable cut resistance with a 13-gauge Tuffalene® UHMWPE shell. A black foam nitrile palm dip provides excellent wet/oily grip and improved abrasion resistance.
  • ANSI level A4 cut resistance
  • EN 388 levels: 4X43D
  • Features a 13-gauge seamless shell
  • Foam nitrile coated palm for high grip
  • Excellent abrasion resistance
  • Knit wrist for secure fit and comfort
  • Suitable for sharp object handling, sheet metal work, and glass handling