Learn More
MSC Global Glove Cut-Resistant Gloves, ANSI A4, Nitrile Coated, 1 Pair
Catalog No. 5000768868
Encompass
These cut-resistant gloves are designed for tasks requiring high dexterity and protection. They feature an 18 gauge seamless knit liner and a nitrile coating for enhanced grip and durability, meeting ANSI A4 cut resistance standards.
  • Seamless knit construction for comfort and dexterity
  • Nitrile coating on palm and fingers for improved grip
  • Reinforced thumb crotch for extended wear
  • Made with Tuffalene HDPE material
  • Meets ANSI A4 and EN 388 4543D cut resistance standards
  • Knit wrist cuff for a secure fit
Catalog No. 50-007-68868 Supplier MSC Supplier No. 74555327
These cut-resistant gloves are designed for tasks requiring high dexterity and protection. They feature an 18 gauge seamless knit liner and a nitrile coating for enhanced grip and durability, meeting ANSI A4 cut resistance standards.
  • Seamless knit construction for comfort and dexterity
  • Nitrile coating on palm and fingers for improved grip
  • Reinforced thumb crotch for extended wear
  • Made with Tuffalene HDPE material
  • Meets ANSI A4 and EN 388 4543D cut resistance standards
  • Knit wrist cuff for a secure fit