Learn More
System Biosciences LLC GLOBAL ULTRARAPID LENTIVIRAL TITER KITUltraRapid Lentiviral Global Titering Kit (Human and Mouse compatible) = 42 Reactions
SDP
Catalog No. NC0325399
Encompass_Preferred
Global UltraRapid™ Lentiviral Titering Kit - Rapidly determine pseudovirus titers by measuring the copy number of integrated lentiviral constructs. With a fast and simple workflow, the Global UltraRapid™ Lentiviral Titering Kit, delivers critical lentiviral titer information so you can move more quickly to the next step, whether that’s transduction of target cells in your own lab or delivery of packaged virus particles to another lab.
Catalog No. NC0325399 Supplier System Biosciences LLC Supplier No. LV961A1
Global UltraRapid™ Lentiviral Titering Kit - Rapidly determine pseudovirus titers by measuring the copy number of integrated lentiviral constructs. With a fast and simple workflow, the Global UltraRapid™ Lentiviral Titering Kit, delivers critical lentiviral titer information so you can move more quickly to the next step, whether that’s transduction of target cells in your own lab or delivery of packaged virus particles to another lab.