Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ GlobalMark™ OSHA CAUTION Sign

Catalog No. 19109009 Shop All Brady Corporation Products
Change view
Click to view available options
CAUTION Sign header
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-109-009 CAUTION Sign header
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-109-009 Supplier Brady™ Supplier No. 121560
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Communicates important safety information and machine operational instructions. Brady Sign Headers are made of Indoor/outdoor grade material and conforms to rough, highly textured, and difficult to adhere to surfaces

  • Made of B-595 vinyl film with permanent adhesive
  • Available in size: 4 x 6.25 in.

Specifications

Adhesive Type Acrylic, Permanent, Cold-temperature
Color Black, Yellow
Finish Gloss
Format Cartridge
Material Vinyl (B-595)
Product Type CAUTION Sign
For Use With (Equipment) GlobalMark Label Maker
Quantity 1 Ea.
Shape Rectangle
Size (English) 6.25 x 4 in. (W x H)
Size (Metric) 158.7 x 101.6 mm (W x H)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.