Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

High-Tech Conversions GlobalTech 99% Isopropyl Alcohol IPA

Catalog No. 17195181
Change view
Click to view available options
Quantity:
16 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-195-181 16 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-195-181 Supplier High-Tech Conversions Supplier No. SB6IPA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Highly effective universal cleaner, degreaser and disinfectant.

  • Removes most types of inks, pastes, fluxes, light oils and other contaminates associated with assembly, machining, soldering and printing.
  • Ideal for use in surface preparation prior to manufacturing or assembly processes.
  • Dries spot free
  • Drying agent
  • Extremely economical
  • Flux thinner
  • Low toxicity
  • Mild odor
  • Non-aggressive
  • Plastic safe
  • Ultra-pure for critical electronic cleaning
  • Very fast evaporator

Specifications

Product Type Isopropyl Alcohol
Form Spray Bottle
Scent Unscented
Sterility Non-sterile
Disposable Single-use
Quantity 16 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.