Learn More
Globe Scientific 5 oz. Specimen Containers
Catalog No. 21200293 Shop All Globe Scientific Products
Change view
Click to view available options
No. per Case:
300/Cs.
Catalog No. No. per Case
21-200-293 300/Cs.
1 options
Catalog No. 21-200-293 Supplier Globe Scientific Supplier No. 5917
Add to Cart
Add to Cart
With leak-resistant snap caps
  • Polypropylene containers with polyethylene caps
  • 70mm wide opening and writing area on lid
  • Available with molded graduation marks
Specifications
No. per Case 300/Cs.