Learn More
Globe Scientific Coagulation Cup for Accu Coa Data Coagulation Analyzers
Catalog No. 21200242 Shop All Globe Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Accu Coa Data Coagulation Analyzers
Catalog No. For Use With (Equipment)
21-200-242 Accu Coa Data Coagulation Analyzers
1 options
Catalog No. 21-200-242 Supplier Globe Scientific Supplier No. 5574
Add to Cart
Add to Cart
For use on coagulation instruments.
  • Made of Polystyrene
  • Supplied with metal mixing bars
  • Dimensions: 1.3 x 0.3 in. (35.1 x 8.6mm)
Specifications
Type Coagulation Cup
For Use With (Equipment) Accu Coa Data Coagulation Analyzers
Includes Metal mixing bars