Learn More
Globe Scientific Needle Holders with Safety Shield
Catalog No. 22171602 Shop All Globe Scientific Products
Change view
Click to view available options
Product Type:
Needle Holder
Catalog No. Product Type
22-171-602 Needle Holder
1 options
This item is not returnable. View return policy
Catalog No. 22-171-602 Supplier Globe Scientific Supplier No. 1204
Add to Cart
Add to Cart
This item is not returnable. View return policy
Designed to protect blood collection personnel from accidental needle sticks
  • Needle is secured by an audible “click” and ready for disposal
  • Fits 13 and 16mm blood collection tubes and all common needle units
  • Complies with OSHA directives by preventing removal of needle and re-use of holder
Specifications
Product Type Needle Holder
Certifications/Compliance OSHA
Disposable Single-use
For Use With (Application) Designed to protect Blood collection personnel from accidental needle sticks
For Use With (Equipment) 13 and 16 mm Blood Collection Tubes and all Common Needle Units
Includes Plastic Safety Shield