Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific Quick-Read Urinalysis Test Slide

Catalog No. 1437759 Shop All Globe Scientific Products
Change view
Click to view available options
Volume (Metric):
6.3μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-377-59 6.3μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-377-59 Supplier Globe Scientific Supplier No. 3805
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Utilizes pre-calibrated circles in the viewing chambers to accurately count cellular elements in urine sediment

  • Combination slide and coverslip of optically clear acrylic with ten separate chambers, each holding a standard volume of 6.3μL
  • Ten processed and stained specimens can be read at a time
  • Each chamber contains 18 premeasured circles that contain specific volumes of urinary sediment within its circumference
  • Chambers facilitate a convenient and rapid microscopic examination of the cellular elements in each specimen
  • Capillary action provides uniform distribution of sediment into the viewing chamber
  • Each chamber has overflow wells for excess urine to prevent cross contamination
  • Individual slides and specimens can be identified by marking the appropriate numbers on the extreme left and right side of the unit

Specifications

Product Type Quick-Read Urinalysis Slide
Material Acrylic
Color Clear
For Use With (Application) Used to accurately count cellular elements in Urine sediment, for microscopic examination
Includes 18 Premeasured counting circles and coverslip
Volume (Metric) 6.3μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.