Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific Sedi-Rate™ ESR System Starter Kit

Catalog No. 21200213 Shop All Globe Scientific Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-200-213 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-200-213 Supplier Globe Scientific Supplier No. 3467
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

100 Sedi-Rate pipettes and vials, 10-place rack with pipette support clamps, leveling platform, 100 piercing funnels and 100 ESR transfer pipettes.

For Westergren ESR determination

  • Closed system for optimum safety
  • Produces accurate and reproducible results
  • Supplied with Polystyrene, graduated pipet with fibrous plug located at zero mark
  • Contains Polypropylene vial pre-filled with 0.25mL of 3.8% trisodium citrate diluent
  • Eliminates splashing, spraying and aerosol hazards

Specifications

Type Sedi-Rate™ Westergren Starter Kit
For Use With (Application) Westergren ESR determination
Includes 100 Sedi-Rate™ Polystyrene graduated pipettes with fibrous plug, Polypropylene vial pre-filled with 0.25mL of 3.8% Trisodium Citrate diluent, 10-place rack with pipette support clamps, leveling platform, 100 piercing funnels and 100 ESR
Quantity 100 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.