Learn More
MSC Globe Scientific Wash Bottle, Wide Mouth, 250 mL, LDPE, Safety Vented
Catalog No. 5000531260
Encompass
This 250 mL wide mouth wash bottle is made from LDPE and features an advanced DripLok venting system to prevent pressure build-up, making it safe for dispensing volatile solvents. It includes blank write-on panels for customizable labeling and indelible safety printing in multiple languages for global compliance.
  • Blank write-on panels for customizable labeling
  • Indelible safety printing in English, French, and German for global understanding
  • Advanced DripLok venting system reduces pressure build-up for safe dispensing of volatile solvents
  • Includes adhesive hazard labels for chemical identification
  • Ideal for laboratories handling multiple solvents and reagents
Catalog No. 50-005-31260 Supplier MSC Supplier No. 60890985
This 250 mL wide mouth wash bottle is made from LDPE and features an advanced DripLok venting system to prevent pressure build-up, making it safe for dispensing volatile solvents. It includes blank write-on panels for customizable labeling and indelible safety printing in multiple languages for global compliance.
  • Blank write-on panels for customizable labeling
  • Indelible safety printing in English, French, and German for global understanding
  • Advanced DripLok venting system reduces pressure build-up for safe dispensing of volatile solvents
  • Includes adhesive hazard labels for chemical identification
  • Ideal for laboratories handling multiple solvents and reagents