Learn More
AdipoGen Globosuxanthone A
SDP
Catalog No. 502562642
Encompass_Preferred
Chemical. CAS 917091-74-8. Formula C15H12O7. MW 304.3. Isolated from fungus species unknown. Potent anticancer agent. Inhibits cell growth, proliferation and survival of human tumor cells lines in vivo and in vitro. Exhibits strong cytotoxicity against a panel of human solid tumor cell lines, disrupting the cell cycle leading to the accumulation of cells in either G2/M or S phase and inducing classic signs of apoptosis. Antifungal agent against Candida albicans.
Catalog No. 50-256-2642 Supplier AdipoGen Supplier No. AGCN20174M001
Chemical. CAS 917091-74-8. Formula C15H12O7. MW 304.3. Isolated from fungus species unknown. Potent anticancer agent. Inhibits cell growth, proliferation and survival of human tumor cells lines in vivo and in vitro. Exhibits strong cytotoxicity against a panel of human solid tumor cell lines, disrupting the cell cycle leading to the accumulation of cells in either G2/M or S phase and inducing classic signs of apoptosis. Antifungal agent against Candida albicans.