Learn More
Cayman Chemical Globotriaosylceramide (porcine RBC), Purity: ≥98%, CAS: 71965-57-6, 10 mg
SDP
Catalog No. 503015868
Encompass_Preferred
A sphingolipid; found in mammalian cell membranes; act as receptors for Shiga and Shiga-like toxins in vitro and in vivo; accumulate in various cells in patients with Fabry disease; interacts with HIV gp120 to prevent its interaction with chemokine co-receptors and subsequent fusion of HIV to host cell membranes
Catalog No. 50-301-5868 Supplier Cayman Chemical Supplier No. 2487010 MG
A sphingolipid; found in mammalian cell membranes; act as receptors for Shiga and Shiga-like toxins in vitro and in vivo; accumulate in various cells in patients with Fabry disease; interacts with HIV gp120 to prevent its interaction with chemokine co-receptors and subsequent fusion of HIV to host cell membranes