Learn More
Crescent Chemical Co Inc GLOBULIN BOVINE
SDP
Supplier:  Crescent Chemical Co Inc SE22550.01
Encompass_Preferred
Globulin bovine Cohn-Fraction II. Highly purified. Soluble in saline and standard buffers. Suitable to reduce non-specific adsorption of antibody in immunoassay systems. Starting material for the isolation of IgG subclasses. Size - 5G Storage Conditions -
Catalog No. 50-247-631