Learn More
Aviva Systems Biology Corp GLOD4 Antibody - N-terminal region (ARP56828_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56828_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GLOD4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human GLOD4. Alternate names: C17orf25, CGI-150, HC71ApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-286-220