Learn More
Agilent Technologies GLOGOS II AUTORAD MARKERS
Catalog No. 50125805
Encompass
Glogos II Autorad Markers,luminescent peel-off stickers for marking and aligning autoradiographs,easily affixed to plastic wrap or paper,convenient, nonhazardous alternative to messy radioactive ink, adaptable for long or short exposures.
Catalog No. 50-125-805 Supplier Agilent Technologies Supplier No. 420203
Glogos II Autorad Markers,luminescent peel-off stickers for marking and aligning autoradiographs,easily affixed to plastic wrap or paper,convenient, nonhazardous alternative to messy radioactive ink, adaptable for long or short exposures.