Learn More
Agilent Technologies Glogos II Autorad Markers, 100 markers
Catalog No. 50125803
Encompass
Glogos II Autorad Markers,luminescent peel-off stickers for marking and aligning autoradiographs,easily affixed to plastic wrap or paper,convenient, nonhazardous alternative to messy radioactive ink, adaptable for long or short exposures. 100 markers
Catalog No. 50-125-803 Supplier Agilent Technologies Supplier No. 420201
Glogos II Autorad Markers,luminescent peel-off stickers for marking and aligning autoradiographs,easily affixed to plastic wrap or paper,convenient, nonhazardous alternative to messy radioactive ink, adaptable for long or short exposures. 100 markers